Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

Skip To

BDPH9BCDED1E

Bide Pharmatech

((2-((5-Fluoro-4-(4-fluoro-2-methoxyphenyl)pyridin-2-yl)amino)pyridin-4-yl)methyl)(imino)(methyl)-l6-sulfanone

Synonym(s):

1-(2-((5-fluoro-4-(4-fluoro-2-methoxyphenyl)pyridin-2-yl)amino)pyridin-4-yl)-S-methylmethanesulfinamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C19H18O2N4S1F2
CAS Number:
Molecular Weight:
404.44
MDL number:
Product BDPH9BCDED1E is currently not available for purchase. Contact Technical Service

Inchi Code

1S/C19H18F2N4O2S/c1-27-17-8-13(20)3-4-14(17)15-9-19(24-10-16(15)21)25-18-7-12(5-6-23-18)11-28(2,22)26/h3-11H,1-2H3,(H2,22,26)(H,23,24,25)

Inchi Key

PFBNXCPSFMJHFF-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library