Skip to Content
MilliporeSigma

BDPH9BCDFD32

Bide Pharmatech

Methyl 1-methyl-1H-indole-4-carboxylate

Synonym(s):

methyl 1-methyl-1H-indole-4-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCDFD32 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C11H11O2N1
CAS Number:
Molecular Weight:
189.21
MDL number:

Skip To

Inchi Code

1S/C11H11NO2/c1-12-7-6-8-9(11(13)14-2)4-3-5-10(8)12/h3-7H,1-2H3

Inchi Key

HVQOOSBDNRKUDW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library