FLUH99C8E395
Fluorochem (Preferred Partner)
2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Sign Into View Organizational & Contract Pricing
Select a Size
Product FLUH99C8E395 is currently not available for purchase. Contact Technical Service
Skip To
Iupac Name
2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Inchi Code
1S/C12H17BClNO2/c1-11(2)12(3,4)17-13(16-11)8-5-6-10(15)9(14)7-8/h5-7H,15H2,1-4H3
Inchi Key
BEXOYZKPTUJSCZ-UHFFFAOYSA-N
Storage temp.
Ambient Storage
Purity
95%
Country of Origin
GB
Physical Form
Solid
Product Link
https://fluorochem.co.uk/product/F338201
Signal Word
Warning
Hazard Statements
MSDS
https://sds.fluorochem.co.uk/eng/sds_F338201_en.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
