Skip to Content
MilliporeSigma

17618

2-Bromomethyl-1,3-dioxolane

purum, ≥95.5% (GC)

Synonym(s):

Bromoacetaldehyde ethylene acetal

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C4H7BrO2
CAS Number:
Molecular Weight:
167.00
EC Number:
224-443-6
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
103309
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI key

CKIIJIDEWWXQEA-UHFFFAOYSA-N

InChI

1S/C4H7BrO2/c5-3-4-6-1-2-7-4/h4H,1-3H2

SMILES string

BrCC1OCCO1

grade

purum

assay

≥95.5% (GC)

refractive index

n20/D 1.482

bp

80-82 °C/27 mmHg (lit.)

density

1.613 g/mL at 25 °C (lit.)

Application

2-Bromomethyl-1,3-dioxolane was used in the synthesis of 1,4-benzoxazepine (BZO) derivatives[1]. It was also used in the preparation of Grignard reagent[2].

Other Notes

Useful synthon for the preparation of α,β-unsaturated aldehydes[3]; Synthon, for the preparation of 1,4-dialdehyde monoacetals[4]

comparable product

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

143.6 °F - closed cup

flash_point_c

62 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.F. Borgne, et al. et al.
Tetrahedron Letters, 1379-1379 (1976)
T.M. Cresp et al.
Journal of the Chemical Society. Perkin Transactions 1, 39-39 (1974)
Schmeichel M and Redlich H.
Synthesis, 1996(08), 1002-1006 (1996)
K Kamei et al.
Bioorganic & medicinal chemistry letters, 11(4), 595-598 (2001-03-07)
A series of new 3-substituted-4-(4-aminobutyl)-1,4-benzoxazepin-5(4H)-one derivatives (1-5) which showed a very high affinity for 5-HT1A receptor with good selectivity over dopamine D2 receptor was synthesized. Among these compounds, 3-chloro-4-[4-[4-(2-pyridinyl)-1,2,3,6-tetrahydropyridin-1-yl]butyl]-1,4-benzoxazepin-5(4H)-one (5: SUN N4057) exhibited remarkable neuroprotective activity in a transient middle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service