MilliporeSigma
  • Mineralization of individual congeners of linear alkylbenzenesulfonate by defined pairs of heterotrophic bacteria.
Mineralization of individual congeners of linear alkylbenzenesulfonate by defined pairs of heterotrophic bacteria.
Applied and environmental microbiology (2004-07-09)
David Schleheck, Thomas P Knepper, Karin Fischer, Alasdair M Cook
ABSTRACT
Parvibaculum lavamentivorans DS-1(T) utilized the commercial surfactant linear alkylbenzenesulfonate (LAS) (20 congeners with C(10) to C(13) side chains) as a carbon and energy source by shortening the side chain, and sulfophenylcarboxylates (SPCs) and similar compounds (e.g., alpha,beta-unsaturated SPCs [SPC-2Hs]) were excreted with quantitative recovery of the sulfophenyl moiety. 2-(4-Sulfophenyl)decane (2-C10-LAS) was converted largely to 3-(4-sulfophenyl)butyrate (3-C4-SPC), as were 2-C12-LAS and 2-C14-LAS; the other products were 5-C6-SPC (SPC+2C) and 3-C4-SPC-2H. 2-C11-LAS was converted largely to 4-C5-SPC with the corresponding SPC+2C and SPC-2H; similarly, 3-C12-LAS yielded 4-C6-SPC with the corresponding SPC+2C and SPC-2H. This pattern of products confirmed that LAS is degraded by omega-oxygenation and chain shortening through beta-oxidation. At least nine major SPCs were formed from commercial LAS. The novel isolates Comamonas testosteroni SPB-2 and KF-1 utilized 3-C4-SPC; Delftia acidovorans SPH-1 utilized 4-C6-SPC enantioselectively. The substrate-dependent oxygen uptake of whole cells of strain SPB-2 indicated that there was inducible oxygenation of 3-C4-SPC and of 4-sulfophenol in whole cells of the strains of C. testosteroni during growth with 3-C4-SPC or 4-sulfophenol. The degradative pathways apparently involved 4-sulfocatechol and 4-sulfocatechol 1,2-dioxygenase. Strain SPB-2 and strain DS-1(T) grew together in LAS-salts medium, and only seven of the nine major SPCs were recovered. Strain SPB-2 utilized 3-C4-SPC, 3-C5-SPC, and 3-C4-SPC-2H. Strain SPH-1 grew together with strain DS-1(T) in LAS-salts medium, and a different set of seven major SPCs was recovered. Strain SPH-1 utilized 4-C6-SPC, 4-C5-SPC, 4-C6-SPC-2H, and 4-C5-SPC-2H. A three-member community consisting of strains DS-1(T), SPB-2, and SPH-1 utilized four major SPCs. We inferred that this community mineralized the major SPCs derived from 8 of the 20 LAS congeners.
Product Description
Supelco
SPB®-1 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 1.00 μm
Pricing and availability is not currently available.