MilliporeSigma
  • The effect of aging on mitochondrial and cytosolic hepatic intrinsic death pathway and apoptosis associated proteins in Fischer 344 rats.
The effect of aging on mitochondrial and cytosolic hepatic intrinsic death pathway and apoptosis associated proteins in Fischer 344 rats.
Experimental gerontology (2015-04-26)
John Mach, Aniko Huizer-Pajkos, Alice Kane, Brett Jones, Catriona McKenzie, Sarah J Mitchell, Rafael de Cabo, Victoria C Cogger, David G Le Couteur, Sarah N Hilmer
ABSTRACT
Apoptosis is increased in the liver in old age and is a common pathological feature of liver disease. The mitochondria play a key role in regulating apoptosis via the intrinsic death pathway. As the effect of aging on this pathway is unclear, we aimed to characterize the impact of aging on the hepatic intrinsic death pathway and apoptosis. Livers from young adult (6.6 ± 0.3 months, n = 9) and old (25.4 ± 0.7 months, n = 9) male Fischer 344 rats were extracted for cellular fractionation and immunobloting. In old age there were lower mitochondrial protein levels of pro-apoptotic BAK, BID, tBID and VDAC1 (p < 0.05) and of anti-apoptotic Bcl-2. Compared to young, old rats had lower cytosolic protein levels of pro-apoptotic BAX, BAK, BID, tBID and anti-apoptotic Bcl-xL (p < 0.05). BAK, Bcl-2 and Bcl-xL were found in the cytosol. Furthermore with old age, cytosolic protein levels of cytochrome C, AIF and cleaved caspase-9 did not change but activation of caspase-3, -6 and -7 increased (p < 0.05) and DNA fragmentation trended to increase. Our results suggest an age-related decline in the levels of a number of proteins involved in the intrinsic death pathway, an uncoupling of intermediate apoptosis signaling and increased cellular apoptosis in the liver in old age.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution