MilliporeSigma
  • CLAP, a novel caspase recruitment domain-containing protein in the tumor necrosis factor receptor pathway, regulates NF-kappaB activation and apoptosis.
CLAP, a novel caspase recruitment domain-containing protein in the tumor necrosis factor receptor pathway, regulates NF-kappaB activation and apoptosis.
The Journal of biological chemistry (1999-06-11)
S M Srinivasula, M Ahmad, J H Lin, J L Poyet, T Fernandes-Alnemri, P N Tsichlis, E S Alnemri
ABSTRACT
Molecules that regulate NF-kappaB activation play critical roles in apoptosis and inflammation. We describe the cloning of the cellular homolog of the equine herpesvirus-2 protein E10 and show that both proteins regulate apoptosis and NF-kappaB activation. These proteins were found to contain N-terminal caspase-recruitment domains (CARDs) and novel C-terminal domains (CTDs) and were therefore named CLAPs (CARD-like apoptotic proteins). The cellular and viral CLAPs induce apoptosis downstream of caspase-8 by activating the Apaf-1-caspase-9 pathway and activate NF-kappaB by acting upstream of the NF-kappaB-inducing kinase, NIK, and the IkB kinase, IKKalpha. Deletion of either the CARD or the CTD domain inhibits both activities. The CARD domain was found to be important for homo- and heterodimerization of CLAPs. Substitution of the CARD domain with an inducible FKBP12 oligomerization domain produced a molecule that can induce NF-kappaB activation, suggesting that the CARD domain functions as an oligomerization domain, whereas the CTD domain functions as the effector domain in the NF-kappaB activation pathway. Expression of the CARD domain of human CLAP abrogates tumor necrosis factor-alpha-induced NF-kappaB activation, suggesting that cellular CLAP plays an essential role in this pathway of NF-kappaB activation.