MilliporeSigma
  • Capecitabine Efficacy Is Correlated with TYMP and RB1 Expression in PDX Established from Triple-Negative Breast Cancers.
Capecitabine Efficacy Is Correlated with TYMP and RB1 Expression in PDX Established from Triple-Negative Breast Cancers.
Clinical cancer research : an official journal of the American Association for Cancer Research (2018-02-22)
Elisabetta Marangoni, Cécile Laurent, Florence Coussy, Rania El-Botty, Sophie Château-Joubert, Jean-Luc Servely, Ludmilla de Plater, Franck Assayag, Ahmed Dahmani, Elodie Montaudon, Fariba Nemati, Justine Fleury, Sophie Vacher, David Gentien, Audrey Rapinat, Pierre Foidart, Nor Eddine Sounni, Agnès Noel, Anne Vincent-Salomon, Marick Lae, Didier Decaudin, Sergio Roman-Roman, Ivan Bièche, Martine Piccart, Fabien Reyal
ABSTRACT
Purpose: Triple-negative breast cancer (TNBC) patients with residual disease after neoadjuvant chemotherapy have a poor outcome. We developed patient-derived xenografts (PDX) from residual tumors to identify efficient chemotherapies and predictive biomarkers in a context of resistance to anthracyclines- and taxanes-based treatments.Experimental Design: PDX were established from residual tumors of primary breast cancer patients treated in neoadjuvant setting. TNBC PDX were treated by anthracyclines, taxanes, platins, and capecitabine. Predictive biomarkers were identified by transcriptomic and immunohistologic analysis. Downregulation of RB1 was performed by siRNA in a cell line established from a PDX.Results: Residual TNBC PDX were characterized by a high tumor take, a short latency, and a poor prognosis of the corresponding patients. With the exception of BRCA1/2-mutated models, residual PDX were resistant to anthracyclines, taxanes, and platins. Capecitabine, the oral prodrug of 5-FU, was highly efficient in 60% of PDX, with two models showing complete responses. Prior treatment of a responder PDX with 5-FU increased expression of thymidylate synthase and decreased efficacy of capecitabine. Transcriptomic and IHC analyses of 32 TNBC PDX, including both residual tumors and treatment-naïve derived tumors, identified RB1 and TYMP proteins as predictive biomarkers for capecitabine response. Finally, RB1 knockdown in a cell line established from a capecitabine-responder PDX decreased sensitivity to 5-FU treatment.Conclusions: We identified capecitabine as efficient chemotherapy in TNBC PDX models established from residual disease and resistant to anthracyclines, taxanes, and platins. RB1 positivity and high expression of TYMP were significantly associated with capecitabine response. Clin Cancer Res; 24(11); 2605-15. ©2018 AACR.
Sigma-Aldrich
MISSION® esiRNA, targeting human RB1
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-TYMP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.