MilliporeSigma
  • Metabolic engineering of Escherichia coli: increase of NADH availability by overexpressing an NAD+-dependent formate dehydrogenase.
Metabolic engineering of Escherichia coli: increase of NADH availability by overexpressing an NAD+-dependent formate dehydrogenase.
Metabolic engineering (2002)
Berrios-Rivera S J, et al.