MilliporeSigma
Mads and Smads in TGF beta signalling.
Current opinion in cell biology (1998-04-30)
L Attisano, J L Wrana
ABSTRACT
The discovery of the Mothers against dpp (Mad) gene in Drosophila has opened a window on an entirely unique signalling pathway that functions to mediate responses to the tumour growth factor beta (TGF beta) superfamily. This pathway, which is comprised of a family of proteins related to Mad, acts to convey signals directly from TGF beta receptors to the nucleus and is implicated in the pathogenesis of human diseases.
Sigma-Aldrich
SMAD4, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.