MilliporeSigma
  • Evaluation of analytical similarity between trastuzumab biosimilar CT-P6 and reference product using statistical analyses.
Evaluation of analytical similarity between trastuzumab biosimilar CT-P6 and reference product using statistical analyses.
mAbs (2018-02-28)
Jihun Lee, Hyun Ah Kang, Jin Soo Bae, Kyu Dae Kim, Kyoung Hoon Lee, Ki Jung Lim, Min Joo Choo, Shin Jae Chang
ABSTRACT
The evaluation of analytical similarity has been a challenging issue for the biosimilar industry because the number of lots for reference and biosimilar products available at the time of development are limited, whilst measurable quality attributes of target molecule are numerous, which can lead to potential bias or false negative/positive conclusions regarding biosimilarity. Therefore, appropriate statistical analyses are highly desirable to achieve a high level of confidence in the similarity evaluation. A recent guideline for the risk-based statistical approaches recommended by the US Food and Drug Administration provides useful tools to systematically evaluate analytical similarity of biosimilar products compared with reference products. Here, we evaluated analytical similarity of CT-P6, a biosimilar product of trastuzumab, with the reference products (EU-Herceptin® or US-Herceptin®) following these statistical approaches. Various quality attributes of trastuzumab were first ranked based on the clinical impact of each attribute and subsequently adjusted to one of three tiers (Tier 1, Tier 2 and Tier 3) considering the characteristics of the assay, the level of attribute present and the feasibility of statistical analysis. Two biological activities with highest potential clinical impact were evaluated by an equivalent test (Tier 1), and other bioactivities and structural/physicochemical properties relevant to the clinical impact were evaluated by a quality range approach (Tier 2). The attributes with low risk ranking or qualitative assay were evaluated by visual comparison (Tier 3). Analytical similarity assessment analyzed by the three tiers clearly demonstrated that CT-P6 exhibits highly similar structural and physicochemical properties, as well as functional activities, compared with the reference products. There were small differences observed in a few quality attributes between CT-P6 and the reference products, but the differences were very minor, and unlikely to impact on clinical outcome. The recently reported equivalent clinical efficacy of CT-P6 with the reference product further supports that CT-P6 is highly similar compared with the reference product in the view of totality-of-evidence.
Sigma-Aldrich
Anti-Human Kappa Light Chains (Bound and Free)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
HercBridge ELISA Kit
Pricing and availability is not currently available.