MilliporeSigma
  • Immunomodulatory activity of a novel polysaccharide from Lonicera japonica in immunosuppressed mice induced by cyclophosphamide.
Immunomodulatory activity of a novel polysaccharide from Lonicera japonica in immunosuppressed mice induced by cyclophosphamide.
PloS one (2018-10-09)
Xiaonan Zhou, Qun Dong, Xianzhao Kan, Lihong Peng, Xingyu Xu, Yun Fang, Jialiang Yang
ABSTRACT
Lonicera japonica is a typical Chinese herbal medicine. We previously reported a method to isolate polysaccharides from Lonicera japonica (LJP). In this study, we first performed a qualitative analysis of LJP using the Fourier Transform Infrared Spectrometer (FT-IR) and explored the monosaccharide composition of LJP using the pre-column derivatization high performance liquid chromatography (HPLC) method. We then investigated the immunomodulatory function of LJP in cyclophosphamide (CTX)-induced immunosuppressed mouse models. The results showed that LJP had the characteristic absorption of typical polysaccharides consisting of 6 types of monosaccharides. In addition, LJP can increase significantly the organ index, splenic lymphocyte proliferation, macrophage phagocytosis, and natural killer (NK) cell activity in CTX-treated mice. LJP could also restore the levels of serum cytokines interleukin (IL-2), tumor necrosis factor (TNF-α) and Interferon-γ (IFN-γ) in the CTX-treated mice. Finally, the results on measuring the T-lymphocytes subsets of spleen also confirmed LJP-induced immunomodulatory activity in immunosuppressed mice from another perspective. Therefore, LJP could be used as a potential immunomodulatory agent.
Sigma-Aldrich
D-(+)-Fucose, ≥97% (GC)
Pricing and availability is not currently available.