MilliporeSigma
  • Exploring (bio)catalytic activities of structurally characterised Cu(ii) and Mn(iii) complexes: histidine recognition and photocatalytic application of Cu(ii) complex and derived CuO nano-cubes.
Exploring (bio)catalytic activities of structurally characterised Cu(ii) and Mn(iii) complexes: histidine recognition and photocatalytic application of Cu(ii) complex and derived CuO nano-cubes.
Dalton transactions (Cambridge, England : 2003) (2018-09-20)
Babli Kumari, Samapti Kundu, Kajari Ghosh, Mahuya Banerjee, Swapan Kumar Pradhan, Sk Manirul Islam, Paula Brandão, Vítor Félix, Debasis Das
ABSTRACT
Structurally characterised two polymeric complexes of Cu(ii) (3) and Mn(iii) (4) of the ligand L (2-(((2-(phenylamino)ethyl)imino)methyl)phenol) have been explored for catecholase-like activity, fluorescence recognition of histidine and catalytic activity towards coupling of aryl iodides with benzamide, leading to N-arylbenzamides. CuO nano-cubes (NCs) prepared by thermal decomposition of the Cu(ii) complex function as photocatalysts for the degradation of methylene blue. Cube-like morphology of CuO nanocrystal and flake-shaped micrometer order Cu(ii) complex have been established from the field emissive scanning electron microscopy (FESEM) images.
Sigma-Aldrich
N-Phenylethylenediamine, 98%
Pricing and availability is not currently available.