MilliporeSigma
  • An improved HPLC system for the analysis and purification of organic amine radiopharmaceuticals.
An improved HPLC system for the analysis and purification of organic amine radiopharmaceuticals.
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes (1989-01-01)
M J Adam, J R Grierson, S Jivan
ABSTRACT
This paper describes the separation of lipophilic amine radiopharmaceuticals on normal phase silica using a reversed phase type eluant. This class of compound usually gives very poor peak symmetry on reversed phase columns. However, excellent separation of a number of amines of interest to Positron Emission Tomography, such as spiperone and its derivatives, has been achieved on this system.
Sigma-Aldrich
S-(+)-O-Desmethylraclopride hydrobromide, solid, ≥98% (HPLC)
Pricing and availability is not currently available.