MilliporeSigma
  • Substrate recognition and ubiquitination of SCFSkp2/Cks1 ubiquitin-protein isopeptide ligase.
Substrate recognition and ubiquitination of SCFSkp2/Cks1 ubiquitin-protein isopeptide ligase.
The Journal of biological chemistry (2007-04-06)
Shuichan Xu, Mahan Abbasian, Palka Patel, Kristen Jensen-Pergakes, Christian R Lombardo, Brian E Cathers, Weilin Xie, Frank Mercurio, Michele Pagano, David Giegel, Sarah Cox
ABSTRACT
p27, an important cell cycle regulator, blocks the G(1)/S transition in cells by binding and inhibiting Cdk2/cyclin A and Cdk2/cyclin E complexes (Cdk2/E). Ubiquitination and subsequent degradation play a critical role in regulating the levels of p27 during cell cycle progression. Here we provide evidence suggesting that both Cdk2/E and phosphorylation of Thr(187) on p27 are essential for the recognition of p27 by the SCF(Skp2/Cks1) complex, the ubiquitin-protein isopeptide ligase (E3). Cdk2/E provides a high affinity binding site, whereas the phosphorylated Thr(187) provides a low affinity binding site for the Skp2/Cks1 complex. Furthermore, binding of phosphorylated p27/Cdk2/E to the E3 complex showed positive cooperativity. Consistently, p27 is also ubiquitinated in a similarly cooperative manner. In the absence of p27, Cdk2/E and Cks1 increase Skp2 phosphorylation. This phosphorylation enhances Skp2 auto-ubiquitination, whereas p27 inhibits both phosphorylation and auto-ubiquitination of Skp2.
Sigma-Aldrich
Anti-phospho-Kip1 (Thr187) Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.