MilliporeSigma
  • The production of poly-(gamma-glutamic acid) from microorganisms and its various applications.
The production of poly-(gamma-glutamic acid) from microorganisms and its various applications.
Bioresource technology (2001-08-14)
I L Shih, Y T Van
ABSTRACT
This review article deals with the chemistry and biosynthesis of poly-(gamma-glutamic acid) (gamma-PGA) produced by various strains of Bacillus. Potential applications of gamma-PGA as thickener, cryoprotectant, humectant, drug carrier, biological adhesive, flocculant, or heavy metal absorbent, etc. with biodegradability in the fields of food, cosmetics, medicine and water treatments are also reviewed.
Sigma-Aldrich
Poly-L-γ-glutamic acid sodium salt
Pricing and availability is not currently available.