MilliporeSigma
Colipase: structure and interaction with pancreatic lipase.
Biochimica et biophysica acta (1999-11-26)
H van Tilbeurgh, S Bezzine, C Cambillau, R Verger, F Carrière
ABSTRACT
Colipase is a small protein cofactor needed by pancreatic lipase for the efficient dietary lipid hydrolysis. It binds to the C-terminal, non-catalytic domain of lipase, thereby stabilising an active conformation and considerably increasing the overall hydrophobic binding site. Structural studies of the complex and of colipase alone have clearly revealed the functionality of its architecture. Interestingly, a structural analogy has recently been discovered between colipase and a domain in a developmental protein (Dickkopf), based on sequence analogy and homology modeling. Whether this structural analogy implies a common function (lipid interaction) remains to be clarified. Structural analogies have also been recognised between the pancreatic lipase C-terminal domain, the N-terminal domains of lipoxygenases and the C-terminal domain of alpha-toxin. These non-catalytic domains in the latter enzymes are important for interaction with membranes. It has not been established if these domains are also involved in eventual protein cofactor binding as is the case for pancreatic lipase.
Sigma-Aldrich
Colipase from porcine pancreas, essentially salt-free, lyophilized powder
Pricing and availability is not currently available.