MilliporeSigma
  • Thrombin and prothrombin are expressed by neurons and glial cells and accumulate in neurofibrillary tangles in Alzheimer disease brain
Thrombin and prothrombin are expressed by neurons and glial cells and accumulate in neurofibrillary tangles in Alzheimer disease brain
Journal of Neuropathology and Experimental Neurology (2006)
Arai T, et al.