MilliporeSigma
  • Carica papaya lipase: a novel biocatalyst for the enantioselective hydrolysis of (R, S)-naproxen 2, 2, 2-trifluoroethyl ester
Carica papaya lipase: a novel biocatalyst for the enantioselective hydrolysis of (R, S)-naproxen 2, 2, 2-trifluoroethyl ester
Enzyme and Microbial Technology (2005)
Chen CC and Tsai SW