MilliporeSigma
  • Inhibition of cGMP-dependent protein kinase by (Rp)-guanosine 3',5'-monophosphorothioates.
Inhibition of cGMP-dependent protein kinase by (Rp)-guanosine 3',5'-monophosphorothioates.
FEBS letters (1990-04-09)
E Butt, M van Bemmelen, L Fischer, U Walter, B Jastorff
ABSTRACT
The activation of the cGMP-dependent protein kinase and cAMP-dependent protein kinase by the diastereomers of guanosine 3',5'-monophosphorothioate, (Sp)-cGMPS and (Rp)-cGMPS, and 8-chloroguanosine 3',5'-monophosphorothioate, (Sp)-8-Cl-cGMPS and (Rp)-8-Cl-cGMPS, was investigated using the peptide Kemptide as substrate. The (Sp)-diastereomers, which have an axial exocyclic sulfur atom, bound to the cGMP-dependent protein kinase and stimulated its phosphotransferase activity. In contrast, the (Rp)-isomers, which have an equatorial exocyclic sulfur atom, bound to the enzyme without stimulation of its activity. (Rp)-cGMPS and (Rp)-8-Cl-cGMPS antagonized the activation of the cGMP-dependent protein kinase with a Ki of 20 microM and 1.5 microM, respectively. (Rp)-cGMPS also antagonized the activation of cAMP-dependent protein kinase with a Ki of 20 microM. In contrast, (Rp)-8-cGMPS ws a weak inhibitor of the cAMP-dependent protein kinase with a Ki of 100 microM. (Rp)-8-Cl-cGMPS appears to be a rather selective inhibitor of the cGMP-dependent protein kinase and may be a useful tool for studying the role of cGMP in broken and intact cell systems.