MilliporeSigma
  • The Fusarium virguliforme toxin FvTox1 causes foliar sudden death syndrome-like symptoms in soybean.
The Fusarium virguliforme toxin FvTox1 causes foliar sudden death syndrome-like symptoms in soybean.
Molecular plant-microbe interactions : MPMI (2011-06-04)
Hargeet K Brar, Sivakumar Swaminathan, Madan K Bhattacharyya
ABSTRACT
Fusarium virguliforme causes sudden death syndrome (SDS) in soybean. The pathogen has never been isolated from diseased foliar tissues; therefore, one or more toxins have been considered to cause foliar SDS development. Cell-free F. virguliforme culture filtrates containing a toxin causes foliar SDS in soybean. A low-molecular-weight protein of approximately 13.5 kDa (FvTox1), purified from F. virguliforme culture filtrates, produces foliar SDS-like symptoms in cut soybean seedlings. Anti-FvTox1 monoclonal antibodies raised against the purified FvTox1 were used in isolating the FvTox1 gene. In the presence of light, recombinant FvTox1 protein expressed in an insect cell line resulted in chlorosis and necrosis in soybean leaf disks that are typical foliar SDS symptoms. SDS-susceptible but not the SDS-resistant soybean lines were sensitive to the baculovirus-expressed toxin. The requirement of light for foliar SDS-like symptom development indicates that FvTox1 induces foliar SDS in soybean, most likely through production of free radicals by interrupting photosynthesis.
Sigma-Aldrich
Sephacryl®, 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans)
Pricing and availability is not currently available.