MilliporeSigma
  • Structure of the plastic-degrading Ideonella sakaiensis MHETase bound to a substrate.
Structure of the plastic-degrading Ideonella sakaiensis MHETase bound to a substrate.
Nature communications (2019-04-14)
Gottfried J Palm, Lukas Reisky, Dominique Böttcher, Henrik Müller, Emil A P Michels, Miriam C Walczak, Leona Berndt, Manfred S Weiss, Uwe T Bornscheuer, Gert Weber
ABSTRACT
The extreme durability of polyethylene terephthalate (PET) debris has rendered it a long-term environmental burden. At the same time, current recycling efforts still lack sustainability. Two recently discovered bacterial enzymes that specifically degrade PET represent a promising solution. First, Ideonella sakaiensis PETase, a structurally well-characterized consensus α/β-hydrolase fold enzyme, converts PET to mono-(2-hydroxyethyl) terephthalate (MHET). MHETase, the second key enzyme, hydrolyzes MHET to the PET educts terephthalate and ethylene glycol. Here, we report the crystal structures of active ligand-free MHETase and MHETase bound to a nonhydrolyzable MHET analog. MHETase, which is reminiscent of feruloyl esterases, possesses a classic α/β-hydrolase domain and a lid domain conferring substrate specificity. In the light of structure-based mapping of the active site, activity assays, mutagenesis studies and a first structure-guided alteration of substrate specificity towards bis-(2-hydroxyethyl) terephthalate (BHET) reported here, we anticipate MHETase to be a valuable resource to further advance enzymatic plastic degradation.