MilliporeSigma
  • The high resolution crystal structure of yeast hexokinase PII with the correct primary sequence provides new insights into its mechanism of action
The high resolution crystal structure of yeast hexokinase PII with the correct primary sequence provides new insights into its mechanism of action
Test (2000)
Kuser PR, et al.