MilliporeSigma
  • Electrospray Film Deposition for Solvent-Elimination Infrared Spectroscopy.
Electrospray Film Deposition for Solvent-Elimination Infrared Spectroscopy.
Applied spectroscopy (2019-04-17)
Andrea Arangio, Christophe Delval, Giulia Ruggeri, Nikunj Dudani, Amir Yazdani, Satoshi Takahama
ABSTRACT
The application of electrospray (ES) for quantitative transfer of analytes from solution to an internal reflection element for analysis by attenuated total reflection Fourier transform infrared (ATR FT-IR) spectroscopy has been developed in this work. The ES ATR FT-IR method is evaluated with non-volatile and semi-volatile organic and inorganic compounds dissolved in pure organic solvents or organics in a mixture with water. The technique demonstrates the capability for rapid solvent evaporation from dilute solutions, facilitating the creation of thin films that allow ATR FT-IR to generate transmission-mode-like spectra. Electrospray ATR FT-IR with multiple reflections displays a linear response ( R2 = 0.95-0.99) in absorbance with the deposited mass and instrumental detection limit < 100 ng, which demonstrates potential for quantitative applications. The method is applicable when crystalline substances are present, even though the formation of particles restricts the upper limit of mass loadings relative to substances forming homogeneous films. In addition to the solvent, semi-volatile compounds can evaporate during the ES process; the magnitude of losses will depend on solution composition and temperature.
Sigma-Aldrich
cis-Pinonic acid, 98%
Pricing and availability is not currently available.