MilliporeSigma
  • Zoledronate suppressed angiogenesis and osteogenesis by inhibiting osteoclasts formation and secretion of PDGF-BB.
Zoledronate suppressed angiogenesis and osteogenesis by inhibiting osteoclasts formation and secretion of PDGF-BB.
PloS one (2017-06-09)
Si-Yong Gao, Guang-Sen Zheng, Lin Wang, Yu-Jie Liang, Si-En Zhang, Xiao-Mei Lao, Kan Li, Gui-Qing Liao
ABSTRACT
Bisphosphonates related osteonecrosis of jaw (BRONJ) is a severe complication of systemic BPs administration, the mechanism of which is still unclarified. Recently, platelet-derived growth factor-BB (PDGF-BB) secreted by preosteoclasts was reported to promote angiogenesis and osteogenesis. This study aimed to clarify whether bisphosphonates suppressed preosteoclasts releasing PDGF-BB, and whether the suppression harmed coupling of angiogenesis and osteogenesis, which could contribute to BRONJ manifestation. Zoledronate significantly inhibited osteoclast formation by tartrate-resistant acid phosphatase (TRAP) staining and PDGF-BB secretion tested by ELISA. In line with decreasing secretion of PDGF-BB by preosteoclasts exposed to zoledronate, conditioned medium (CM) from the cells significantly induced less migration of endothelial progenitor cells (EPCs) and mesenchymal stem cells (MSCs) compared to CM from unexposed preosteoclasts. Meanwhile, angiogenic function of EPCs and osteoblastic differentiation of MSCs also declined when culturing with CM from preosteoclasts treated by zoledronate (PZ-CM), evidenced by tube formation assay of EPCs and alkaline phosphatase activity of MSCs. Western blot assay showed that the expression of VEGF in EPCs and OCN, RUNX2 in MSCs declined when culturing with PZ-CM compared to CM from preostoeclasts without exposure of zoledronate. Our study found that zoledronate was able to suppress preosteoclasts releasing PDGF-BB, resulting in suppression of angiogenesis and osteogenesis. Our study may partly contributed to the mechanism of BRONJ.
Sigma-Aldrich
Anti-VEGF Antibody, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.