MilliporeSigma
  • Blocking immunosuppression by human Tregs in vivo with antibodies targeting integrin αVβ8.
Blocking immunosuppression by human Tregs in vivo with antibodies targeting integrin αVβ8.
Proceedings of the National Academy of Sciences of the United States of America (2017-11-08)
Julie Stockis, Stéphanie Liénart, Didier Colau, Amandine Collignon, Stephen L Nishimura, Dean Sheppard, Pierre G Coulie, Sophie Lucas
ABSTRACT
Human regulatory T cells (Tregs) suppress other T cells by converting the latent, inactive form of TGF-β1 into active TGF-β1. In Tregs, TGF-β1 activation requires GARP, a transmembrane protein that binds and presents latent TGF-β1 on the surface of Tregs stimulated through their T cell receptor. However, GARP is not sufficient because transduction of GARP in non-Treg T cells does not induce active TGF-β1 production. RGD-binding integrins were shown to activate TGF-β1 in several non-T cell types. Here we show that αVβ8 dimers are present on stimulated human Tregs but not in other T cells, and that antibodies against αV or β8 subunits block TGF-β1 activation in vitro. We also show that αV and β8 interact with GARP/latent TGF-β1 complexes in human Tregs. Finally, a blocking antibody against β8 inhibited immunosuppression by human Tregs in a model of xenogeneic graft-vs.-host disease induced by the transfer of human T cells in immunodeficient mice. These results show that TGF-β1 activation on the surface of human Tregs implies an interaction between the integrin αVβ8 and GARP/latent TGF-β1 complexes. Immunosuppression by human Tregs can be inhibited by antibodies against GARP or against the integrin β8 subunit. Such antibodies may prove beneficial against cancer or chronic infections.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-TPP1 antibody produced in mouse, clone 3B1, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.