MilliporeSigma
  • Beta-agarases I and II from Pseudomonas atlantica. Substrate specificities.
Beta-agarases I and II from Pseudomonas atlantica. Substrate specificities.
European journal of biochemistry (1983-12-01)
L M Morrice, M W McLean, W F Long, F B Williamson
ABSTRACT
Beta-Agarase I and II were characterised by their action on agar-type polysaccharides and oligosaccharides. Beta-Agarase I, an endo-enzyme, was specific for regions containing a minimum of one unsubstituted neoagarobiose unit [3,6-anhydro-alpha-L-galactopyranosyl-(1 leads to 3)-D-galactose], hydrolysing at the reducing side of this moiety. Yaphe demonstrated that agar was degraded by this enzyme to neoagaro-oligosaccharides limited by the disaccharide but with a predominance of the tetramer [Yaphe, W. (1957) Can. J. Microbiol. 3, 987-993]. Beta-Agarase I slowly degraded neoagarohexaose but not the homologous tetrasaccharide. [1-3H]Neoagarohexaitol was cleaved to neoagarotetraose and [1-3H]neoagarobiitol. The highly substituted agar, porphyran was degraded to methylated, sulphated and unsubstituted neoagaro-oligosaccharides which were invariably terminated at the reducing end by unsubstituted neoagarobiose. The novel enzyme, beta-agarase II, was shown to be an endo-enzyme. Preliminary evidence indicated this enzyme was specific for sequences containing neoagarobiose and/or 6(1)-O-methyl-neoagarobiose. It degraded agar to neoagaro-oligosaccharides of which the disaccharide was limiting and predominant. Beta-Agarase II rapidly degraded isolated neogarotetraose and neoagarohexaose to the disaccharide. With [1-3H]neoagarohexaitol, exo-action was observed, the alditol being cleaved to neoagarobiose and [1-3H]neoagarotetraitol. Neoagarotetraitol was hydrolysed at 4% of the rate observed for the hexaitol. Porphyran was degraded to oligosaccharides, the neutral fraction comprising 24% of the starting carbohydrate. This fraction was almost exclusively disaccharides (22.4%) containing neoagarobiose (7.4%) and 6(1)-O-methyl-neoagarobiose (15%). Beta-Agarase II is probably the 'beta-neoagarotetraose hydrolase' reported by Groleau and Yaphe as an exoenzyme against neoagaro-oligosaccharides [Groleau, D. and Yaphe, W. (1977) Can. J. Microbiol. 23, 672-679].