MilliporeSigma
  • Down-regulation of 5S rRNA by miR-150 and miR-383 enhances c-Myc-rpL11 interaction and inhibits proliferation of esophageal squamous carcinoma cells.
Down-regulation of 5S rRNA by miR-150 and miR-383 enhances c-Myc-rpL11 interaction and inhibits proliferation of esophageal squamous carcinoma cells.
FEBS letters (2015-11-27)
Xinyu Wang, Yanli Ren, Zhiqiong Wang, Xiangyu Xiong, Sichong Han, Wenting Pan, Hongwei Chen, Liqing Zhou, Changchun Zhou, Qipeng Yuan, Ming Yang
ABSTRACT
5S rRNA plays an important part in ribosome biology and is over-expression in multiple cancers. In this study, we found that 5S rRNA is a direct target of miR-150 and miR-383 in esophageal squamous cell carcinoma (ESCC). Overexpression of miR-150 and miR-383 inhibited ESCC cell proliferation in vitro and in vivo. Moreover, 5S rRNA silencing by miR-150 and miR-383 might intensify rpL11-c-Myc interaction, which attenuated role of c-Myc as an oncogenic transcriptional factor and dysregulation of multiple c-Myc target genes. Taken together, our results highlight the involvement of miRNAs in ribosomal regulation during tumorigenesis.
Sigma-Aldrich
Anti-Mdmx Antibody, clone 8C6, clone 8C6, from mouse
Pricing and availability is not currently available.