MilliporeSigma
  • In Vitro Benznidazole and Nifurtimox Susceptibility Profile of Trypanosoma cruzi Strains Belonging to Discrete Typing Units TcI, TcII, and TcV.
In Vitro Benznidazole and Nifurtimox Susceptibility Profile of Trypanosoma cruzi Strains Belonging to Discrete Typing Units TcI, TcII, and TcV.
Pathogens (Basel, Switzerland) (2019-10-23)
Susana Revollo, Bruno Oury, Andrea Vela, Michel Tibayrenc, Denis Sereno
ABSTRACT
We ascertain the in vitro Benznidazole (BZN) and Nifurtimox (NFX) susceptibility pattern of epimastigotes, trypomastigotes, and amastigotes of 21 T. cruzi strains, from patients, reservoir, and triatomine bugs of various geographic origins. Using this panel of isolates, we compute the Epidemiological cut off value (COwt). Then, the frequency of the susceptible phenotype (Wild type) towards benznidazole (BZN) and nifurtimox (NFX) within this set of strains belonging to three discrete typing units (DTUs), TcI, TcII, and TcV, was deduced. We observed that the susceptibility status of individual T. cruzi isolates toward BZN and NFX is related to the genetic background and underlying factors that are probably related to the individual life trait history of each strain. Analyzing drug susceptibility in this conceptual framework would offer the possibility to evidence a link between isolates expressing a low susceptibility level (not wild-type) as defined by the COwt value and none-curative treatment. It will also permit us to track drug-resistant parasites in the T. cruzi population.
Sigma-Aldrich
N-Benzyl-2-nitro-1H-imidazole-1-acetamide, 97%
Pricing and availability is not currently available.