MilliporeSigma
  • Poly(amino acids) as a potent self-adjuvanting delivery system for peptide-based nanovaccines.
Poly(amino acids) as a potent self-adjuvanting delivery system for peptide-based nanovaccines.
Science advances (2020-02-18)
Mariusz Skwarczynski, Guangzu Zhao, Jennifer C Boer, Victoria Ozberk, Armira Azuar, Jazmina Gonzalez Cruz, Ashwini Kumar Giddam, Zeinab G Khalil, Manisha Pandey, Mohini A Shibu, Waleed M Hussein, Reshma J Nevagi, Michael R Batzloff, James W Wells, Robert J Capon, Magdalena Plebanski, Michael F Good, Istvan Toth
ABSTRACT
To be optimally effective, peptide-based vaccines need to be administered with adjuvants. Many currently available adjuvants are toxic, not biodegradable; they invariably invoke adverse reactions, including allergic responses and excessive inflammation. A nontoxic, biodegradable, biocompatible, self-adjuvanting vaccine delivery system is urgently needed. Herein, we report a potent vaccine delivery system fulfilling the above requirements. A peptide antigen was coupled with poly-hydrophobic amino acid sequences serving as self-adjuvanting moieties using solid-phase synthesis, to produce fully defined single molecular entities. Under aqueous conditions, these molecules self-assembled into distinct nanoparticles and chain-like aggregates. Following subcutaneous immunization in mice, these particles successfully induced opsonic epitope-specific antibodies without the need of external adjuvant. Mice immunized with entities bearing 15 leucine residues were able to clear bacterial load from target organs without triggering the release of soluble inflammatory mediators. Thus, we have developed a well-defined and effective self-adjuvanting delivery system for peptide antigens.
Product Description
Millipore
Multiscreen® 96 well Plate, hydrophobic PVDF membrane, pore size 0.45 μm, sterile
Pricing and availability is not currently available.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Pricing and availability is not currently available.