MilliporeSigma
  • Pharmacological properties of AR-C239, 2-[2-[4(o-methoxyphenyl)-piperazine-1-Yl]-ethyl]4,4-dimethyl-1,3(2H-4H) isoquinolinedione, a new alpha-adrenoceptor blocking drug.
Pharmacological properties of AR-C239, 2-[2-[4(o-methoxyphenyl)-piperazine-1-Yl]-ethyl]4,4-dimethyl-1,3(2H-4H) isoquinolinedione, a new alpha-adrenoceptor blocking drug.
Journal of cardiovascular pharmacology (1980-01-01)
P Mouillé, A M Huchet, J Chelly, B Lucet, M F Doursout, H Schmitt
ABSTRACT
In pentobarbital-treated dogs and rats, AR-C239, a new and potent alpha-adrenoceptor blocking drug, competitively antagonized pressor responses to adrenaline and inhibited pressor responses to noradrenaline, phenylephrine, tyramine, and dimethylphenylpiperazinium. Injected intravenously into closed-chest dogs, AR-C239 (3-50 micrograms/kg) induced a progressive fall in blood pressure, heart rate, and sympathetic nerve activity. The drug appears to be devoid of direct vasodilator action, and the fall in blood pressure results from the peripheral alpha-blockade. AR-C239 did not change the tachycardia induced by stimulation of the cardiac nerve in dogs and, at least in this preparation, seems to be a specific alpha 1-adrenoceptor blocking drug. When administered into the cisterna magna of dogs, AR-C239 did not have any centrally mediated cardiovascular actions and heart rate. AR-C239 did not modify the functioning of the baroreflex arc. Due to its specificity for alpha 1-Adrenoceptors, AR-C239 may be useful for characterizing alpha-adrenoceptors.
Sigma-Aldrich
ARC 239 dihydrochloride hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.