MilliporeSigma
  • Expression of heparan sulfate D-glucosaminyl 3-O-sulfotransferase isoforms reveals novel substrate specificities.
Expression of heparan sulfate D-glucosaminyl 3-O-sulfotransferase isoforms reveals novel substrate specificities.
The Journal of biological chemistry (1999-02-13)
J Liu, N W Shworak, P Sinaÿ, J J Schwartz, L Zhang, L M Fritze, R D Rosenberg
ABSTRACT
The 3-O-sulfation of glucosamine residues is an important modification during the biosynthesis of heparan sulfate (HS). Our previous studies have led us to purify and molecularly clone the heparan sulfate D-glucosaminyl 3-O-sulfotransferase (3-OST-1), which is the key enzyme converting nonanticoagulant heparan sulfate (HSinact) to anticoagulant heparan sulfate (HSact). In this study, we expressed and characterized the full-length cDNAs of 3-OST-1 homologous genes, designated as 3-OST-2, 3-OST-3A, and 3-OST-3B as described in the accompanying paper (Shworak, N. W., Liu, J., Petros, L. M., Zhang, L., Kobayashi, M., Copeland, N. G., Jenkins, N. A., and Rosenberg, R. D. (1999) J. Biol. Chem. 274, 5170-5184). All these cDNAs were successfully expressed in COS-7 cells, and heparan sulfate sulfotransferase activities were found in the cell extracts. We demonstrated that 3-OST-2, 3-OST-3A, and 3-OST-3B are heparan sulfate D-glucosaminyl 3-O-sulfotransferases because the enzymes transfer sulfate from adenosine 3'-phosphophate 5'-phospho-[35S]sulfate ([35S]PAPS) to the 3-OH position of glucosamine. 3-OST-3A and 3-OST-3B sulfate an identical disaccharide. HSact conversion activity in the cell extract transfected by 3-OST-1 was shown to be 300-fold greater than that in the cell extracts transfected by 3-OST-2 and 3-OST-3A, suggesting that 3-OST-2 and 3-OST-3A do not make HSact. The results of the disaccharide analysis of the nitrous acid-degraded [35S]HS suggested that 3-OST-2 transfers sulfate to GlcA2S-GlcNS and IdoA2S-GlcNS; 3-OST-3A transfers sulfate to IdoA2S-GlcNS. Our results demonstrate that the 3-O-sulfation of glucosamine is generated by different isoforms depending on the saccharide structures around the modified glucosamine residue. This discovery has provided evidence for a new cellular mechanism for generating a defined saccharide sequence in structurally complex HS polysaccharide.
Sigma-Aldrich
D-Glucosamine 3-sulfate, ≥98.0% (TLC)
Pricing and availability is not currently available.