MilliporeSigma
  • In vitro evaluation of activities of nitazoxanide and tizoxanide against anaerobes and aerobic organisms.
In vitro evaluation of activities of nitazoxanide and tizoxanide against anaerobes and aerobic organisms.
Antimicrobial agents and chemotherapy (1996-10-01)
L Dubreuil, I Houcke, Y Mouton, J F Rossignol
ABSTRACT
The antibacterial activities of nitazoxanide and its main metabolite, tizoxanide, were tested against a broad range of bacteria, including anaerobes. Metronidazole, amoxicillin, amoxicillin-clavulanic acid, piperacillin, cefoxitin, imipenem, and clindamycin were used as positive controls. MICs were determined by reference agar dilution methods. The 241 anaerobes were all inhibited by nitazoxanide, with the MICs at which 90% of isolates are inhibited (MIC90S) being between 0.06 and 4 mg/liter with the exception of those for Propionibacterium species, for which the MIC90 was 16 mg/liter. The MIC90s of nitazoxanide were 0.5 mg/liter for the Bacteroides fragilis group (80 strains), 0.06 mg/liter for Clostridium difficile (21 strains), and 0.5 mg/liter for Clostridium perfringens (16 strains). Metronidazole showed a level of activity comparable to that of nitazoxanide except against Bifidobacterium species, against which it was poorly active, and Propionibacterium species, which were resistant to metronidazole. The other antibiotics showed various levels of activity against anaerobes, with imipenem along with nitazoxanide being the most active agents tested. Tizoxanide was less effective than nitazoxanide except against the B. fragilis group, against which its activity was similar to that of nitazoxanide. Under aerobic conditions, nitazoxanide demonstrated poor activity against members of the family Enterobacteriacae and Pseudomonas, Staphylococcus, and Enterococcus species. The same results were obtained when culture was performed under anaerobic conditions with the notable exception of the results against Staphylococcus aureus. The MICs of nitazoxanide were in the range of 2 to 4 mg/liter for 34 clinical isolates of S. aureus, 12 of which were methicillin resistant, while tizoxanide was not effective.
Sigma-Aldrich
Tizoxanide, ≥98% (HPLC)
Pricing and availability is not currently available.