MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Early 5-HT6 receptor blockade prevents symptom onset in a model of adolescent cannabis abuse.
Early 5-HT6 receptor blockade prevents symptom onset in a model of adolescent cannabis abuse.
EMBO molecular medicine (2020-04-25)
Coralie Berthoux, Al Mahdy Hamieh, Angelina Rogliardo, Emilie L Doucet, Camille Coudert, Fabrice Ango, Katarzyna Grychowska, Séverine Chaumont-Dubel, Pawel Zajdel, Rafael Maldonado, Joël Bockaert, Philippe Marin, Carine Bécamel
ABSTRACT
Cannabis abuse during adolescence confers an increased risk for developing later in life cognitive deficits reminiscent of those observed in schizophrenia, suggesting common pathological mechanisms that remain poorly characterized. In line with previous findings that revealed a role of 5-HT6 receptor-operated mTOR activation in cognitive deficits of rodent developmental models of schizophrenia, we show that chronic administration of ∆9-tetrahydrocannabinol (THC) to mice during adolescence induces a long-lasting activation of mTOR in prefrontal cortex (PFC), alterations of excitatory/inhibitory balance, intrinsic properties of layer V pyramidal neurons, and long-term depression, as well as cognitive deficits in adulthood. All are prevented by administrating a 5-HT6 receptor antagonist or rapamycin, during adolescence. In contrast, they are still present 2 weeks after the same treatments delivered at the adult stage. Collectively, these findings suggest a role of 5-HT6 receptor-operated mTOR signaling in abnormalities of cortical network wiring elicited by THC at a critical period of PFC maturation and highlight the potential of 5-HT6 receptor antagonists as early therapy to prevent cognitive symptom onset in adolescent cannabis abusers.
Sigma-Aldrich
Cesium chloride, Grade I, ≥99.0%
Sigma-Aldrich
ZD7288 hydrate, ≥98% (HPLC)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Sigma-Aldrich
Phosphocreatine disodium salt hydrate, ≥97%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP-conjugate, 1 mg/mL, Upstate®
Sigma-Aldrich
Anti-Glutamate Decarboxylase Antibody, 65 kDa isoform, clone GAD-6, clone GAD-6, Chemicon®, from mouse
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Guanosine 5′-triphosphate sodium salt hydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
CNQX disodium salt hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Anti-Serotonin (5-HT) Transporter (602-622) Rabbit pAb, lyophilized, Calbiochem®
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, HRP conjugate, Upstate®, from goat