MilliporeSigma
  • Intelligent Poly(N-Isopropylmethacrylamide) Hydrogels: Synthesis, Structure Characterization, Stimuli-Responsive Swelling Properties, and Their Radiation Decomposition.
Intelligent Poly(N-Isopropylmethacrylamide) Hydrogels: Synthesis, Structure Characterization, Stimuli-Responsive Swelling Properties, and Their Radiation Decomposition.
Polymers (2020-05-18)
Snežana Ilić-Stojanović, Maja Urošević, Ljubiša Nikolić, Djordje Petrović, Jelena Stanojević, Stevo Najman, Vesna Nikolić
ABSTRACT
Poly(N-isopropylmethacrylamide) (p(NiPMAm)) is one of the lesser known homopolymers that has significant potential for designing new "intelligent" materials. The aims of this work were the synthesis a series of cross-linked p(NiPMAm) hydrogels by the free radical polymerization method and the application of gamma-ray radiation for additional cross-linking. The synthesized p(NiPMAm) hydrogels were structurally characterized by Fourier transform infrared spectroscopy (FTIR) and scanning electron microscopy (SEM). The amount of unreacted monomers was analyzed using high pressure liquid chromatography (HPLC) to evaluate conversion of monomers into polymers. The swelling behavior was monitored in dependence of pH and temperature changes. The previous aim of gamma-ray radiation was the further the cross-linkage of the obtained hydrogel sample in the gelatinous, paste-like state, but the gamma-ray radiation caused decomposition. After absorbing irradiation doses, they transformed into the liquid phase. The results obtained by the gel permeation chromatography (GPC) method indicated that only oligomers and monomers were present in the irradiated liquid material, without molecules with a higher average molar mass, i.e., that the decomposition of the hydrogels occurred. Additionally, the irradiated liquid material was analyzed using the static headspace gas chromatography mass spectrometry (HSS-GC/MS) and gas chromatography/flame ionization detection (HSS-GC/FID) methods. The presence of unchanged initiator molecule and a dominant amount of four new molecules that were different from homopolymers and the reactant (monomer and cross-linker) were determined.
Sigma-Aldrich
N-Isopropylmethacrylamide, 97%
Pricing and availability is not currently available.