MilliporeSigma
  • Leveraging Nanocrystal HKUST-1 in Mixed-Matrix Membranes for Ethylene/Ethane Separation.
Leveraging Nanocrystal HKUST-1 in Mixed-Matrix Membranes for Ethylene/Ethane Separation.
Membranes (2020-04-23)
Chong Yang Chuah, S A S C Samarasinghe, Wen Li, Kunli Goh, Tae-Hyun Bae
ABSTRACT
The energy-intensive ethylene/ethane separation process is a key challenge to the petrochemical industry. HKUST-1, a metal-organic framework (MOF) which possesses high accessible surface area and porosity, is utilized in mixed-matrix membrane fabrication to investigate its potential for improving the performance for C2H4/C2H6 separation. Prior to membrane fabrication and gas permeation analysis, nanocrystal HKUST-1 was first synthesized. This step is critical in order to ensure that defect-free mixed-matrix membranes can be formed. Then, polyimide-based polymers, ODPA-TMPDA and 6FDA-TMPDA, were chosen as the matrices. Our findings revealed that 20 wt% loading of HKUST-1 was capable of improving C2H4 permeability (155% for ODPA-TMPDA and 69% for 6FDA-TMPDA) without excessively sacrificing the C2H4/C2H6 selectivity. The C2H4 and C2H6 diffusivity, as well as solubility, were also improved substantially as compared to the pure polymeric membranes. Overall, our results edge near the upper bound, confirming the effectiveness of leveraging nanocrystal HKUST-1 filler for performance enhancements in mixed-matrix membranes for C2H4/C2H6 separation.
Sigma-Aldrich
2,4,6-Trimethyl-m-phenylenediamine, 96%
Pricing and availability is not currently available.