MilliporeSigma
  • Thermotolerant glycosyl hydrolases-producing Bacillus aerius CMCPS1 and its saccharification efficiency on HCR-laccase (LccH)-pretreated corncob biomass.
Thermotolerant glycosyl hydrolases-producing Bacillus aerius CMCPS1 and its saccharification efficiency on HCR-laccase (LccH)-pretreated corncob biomass.
Biotechnology for biofuels (2020-07-21)
Meena Ganesan, Remitha Mathivani Vinayakamoorthy, Sugitha Thankappan, Iniyakumar Muniraj, Sivakumar Uthandi
ABSTRACT
The current production of bioethanol based on lignocellulosic biomass (LCB) highly depends on thermostable enzymes and extremophiles owing to less risk of contamination. Thermophilic bacterial cellulases are preferred over fungi due to their higher growth rate, presence of complex multi-enzymes, stability, and enhanced bioconversion efficiency. Corncob, underutilized biomass, ensures energy conservation due to high lignocellulosic and more fermentable sugar content. In the present study, the thermophilic bacterium Bacillus aerius CMCPS1, isolated from the thermal springs of Manikaran, Himachal Pradesh, India, was characterized in terms of its activity, stability, and hydrolytic capacity. A two-step process comprising: (i) a combined strategy of hydrodynamic cavitation reaction (HCR)-coupled enzymatic (LccH at 6.5 U) pretreatment for delignification and (ii) subsequent hydrolysis of pre-treated (HCR-LccH) corncob biomass (CCB) using a thermostable cocktail of CMCPS1 was adopted to validate the efficiency of the process. Some of the parameters studied include lignin reduction, cellulose increase, and saccharification efficiency. Among the five isolates obtained by in situ enrichment on various substrates, B. aerius CMCPS1, isolated from hot springs, exhibited the maximum hydrolytic activity of 4.11. The GH activity of the CMCPS1 strain under submerged fermentation revealed maximum filter paper activity (FPA) and endoglucanase activity of 4.36 IU mL-1 and 2.98 IU mL-1, respectively, at 44 h. Similarly, the isolate produced exoglucanase and β-glucosidase with an activity of 1.76 IU mL-1 and 1.23 IU mL-1 at 48 h, respectively. More specifically, the enzyme endo-1,4-β-d glucanase E.C.3.2.1.4 (CMCase) produced by B. aerius CMCPS1 displayed wider stability to pH (3-9) and temperature (30-90 °C) than most fungal cellulases. Similarly, the activity of CMCase increased in the presence of organic solvents (118% at 30% acetone v/v). The partially purified CMCase from the culture supernatant of CMCPS1 registered 64% yield with twofold purification. The zymogram and SDS-PAGE analyses further confirmed the CMCase activity with an apparent molecular mass of 70 kDa. The presence of genes specific to cellulases, such as cellulose-binding domain CelB, confirmed the presence of GH family 46 and β-glucosidase activity (GH3). The multifunctional cellulases of CMCPS1 were evaluated for their saccharification efficiency on laccase (LccH, a fungal laccase from Hexagonia hirta MSF2)-pretreated corncob in a HCR. The lignin and hemicelluloses removal efficiency of HCR-LccH was 54.1 and 6.57%, respectively, with an increase in cellulose fraction (42.25%). The saccharification efficiency of 55% was achieved with CMCPS1 multifunctional cellulases at 50 °C and pH 5.0. The multifunctional cellulase complex of B. aerius CMCPS1 is a potential biocatalyst for application in lignocellulosic biomass-based biorefineries. The saccharification ability of HCR-LccH-pretreated corncob at elevated temperatures would be an advantage for biofuel production from lignocellulosic biomass.
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
Pricing and availability is not currently available.