MilliporeSigma
  • Facile radiosynthesis of fluorine-18 labeled beta-blockers. Synthesis, radiolabeling, and ex vivo biodistribution of [18F]-(2S and 2R)-1-(1-fluoropropan-2-ylamino)-3-(m-tolyloxy)propan-2-ol.
Facile radiosynthesis of fluorine-18 labeled beta-blockers. Synthesis, radiolabeling, and ex vivo biodistribution of [18F]-(2S and 2R)-1-(1-fluoropropan-2-ylamino)-3-(m-tolyloxy)propan-2-ol.
Journal of medicinal chemistry (2008-07-29)
Karin A Stephenson, Alan A Wilson, Jeffrey H Meyer, Sylvain Houle, Neil Vasdev
ABSTRACT
An efficient and general method has been developed for fluorine-18 labeling of beta-blockers that possess the propanolamine moiety. A new synthetically versatile intermediate, 3-(1-(benzyloxy)propan-2-yl)-2-oxooxazolidin-5-yl)methyl 4-methylbenzenesulfonate (13), was prepared and can be conjugated to any phenoxy core. To demonstrate the synthetic methodology, fluorinated derivatives of toliprolol were prepared, namely, [(18)F]-(2S and 2R)-1-(1-fluoropropan-2-ylamino)-3-(m-tolyloxy)propan-2-ol ((2S and 2R)-[(18)F]1). The radiosyntheses were accomplished in <1 h, with 20-24% (uncorrected for decay, n = 7) radiochemical yields, >96% radiochemical and >99% enantiomeric purities, with specific activities of 0.9-1.1 Ci/micromol (EOS). Ex vivo biodistribution studies with the radiotracers demonstrated excessively rapid washout that may limit their use for cerebral PET imaging.
Product Description
Supelco
Carazolol, analytical standard
Pricing and availability is not currently available.