MilliporeSigma
  • γ-Glutamyl transpeptidase is a heavily N-glycosylated heterodimer in HepG2 cells.
γ-Glutamyl transpeptidase is a heavily N-glycosylated heterodimer in HepG2 cells.
Archives of biochemistry and biophysics (2010-09-14)
Matthew B West, Marie H Hanigan
ABSTRACT
The cell surface enzyme γ-glutamyl transpeptidase (GGT) is expressed by human hepatocellular carcinomas (HCCs). HCCs arise from malignant transformation of hepatocytes and are the most common form of primary liver cancer. Identification of tumor-specific, post-translational modifications of GGT may provide novel biomarkers for HCC. The HepG2 cell line, derived from a human HCC, has been used extensively in studies of liver cancer. However, the use of this cell line for studies of GGT have been stymied by reports that HepG2 cells do not process the GGT propeptide into its heterodimeric subunits. The data in this study demonstrate that HepG2 cells do, in fact, produce the mature heterodimeric form of GGT. Immunohistochemical and immunoaffinity analyses provide direct evidence that, in HepG2 cells, GGT is properly localized to the bile canaliculi. Three independent, experimental approaches demonstrate that GGT in HepG2 cells is comprised of two subunits that are more heavily N-glycosylated than GGT from normal human liver tissue. These data directly contradict the dogma in the field. These data support the use of HepG2 cells as a model system for analyzing tumor-specific changes in the post-translational modifications of GGT.
Sigma-Aldrich
L-Glutamic acid γ-(4-methoxy-β-naphthylamide)
Pricing and availability is not currently available.