MilliporeSigma
  • Dynamic bacterial community response to Akashiwo sanguinea (Dinophyceae) bloom in indoor marine microcosms.
Dynamic bacterial community response to Akashiwo sanguinea (Dinophyceae) bloom in indoor marine microcosms.
Scientific reports (2021-03-28)
Seung Won Jung, Junsu Kang, Joon Sang Park, Hyoung Min Joo, Sung-Suk Suh, Donhyug Kang, Taek-Kyun Lee, Hyun-Jung Kim
ABSTRACT
We investigated the dynamics of the bacterial composition and metabolic function within Akashiwo sanguinea bloom using a 100-L indoor microcosm and metagenomic next-generation sequencing. We found that the bacterial community was classified into three groups at 54% similarity. Group I was associated with "during the A. sanguinea bloom stage" and mainly consisted of Alphaproteobacteria, Flavobacteriia and Gammaproteobacteria. Meanwhile, groups II and III were associated with the "late bloom/decline stage to post-bloom stage" with decreased Flavobacteriia and Gammaproteobacteria in these stages. Upon the termination of the A. sanguinea bloom, the concentrations of inorganic nutrients (particularly PO43-, NH4+ and dissolved organic carbon) increased rapidly and then decreased. From the network analysis, we found that the A. sanguinea node is associated with certain bacteria. After the bloom, the specific increases in NH4+ and PO43- nodes are associated with other bacterial taxa. The changes in the functional groups of the bacterial community from chemoheterotrophy to nitrogen association metabolisms were consistent with the environmental impacts during and after A. sanguinea bloom. Consequently, certain bacterial communities and the environments dynamically changed during and after harmful algal blooms and a rapid turnover within the bacterial community and their function can respond to ecological interactions.
Product Description
Millipore
Polycarbonate Membrane Filter, 3.0 μm Pore Size, Isopore, filter diam. 47 mm, hydrophilic
Pricing and availability is not currently available.