MilliporeSigma
  • The 2010 global proficiency study of human papillomavirus genotyping in vaccinology.
The 2010 global proficiency study of human papillomavirus genotyping in vaccinology.
Journal of clinical microbiology (2012-04-27)
Carina Eklund, Ola Forslund, Keng-Ling Wallin, Tiequn Zhou, Joakim Dillner
ABSTRACT
Accurate and internationally comparable human papillomavirus (HPV) DNA genotyping is essential both for evaluation of HPV vaccines and for effective monitoring and implementation of vaccination programs. The World Health Organization (WHO) HPV Laboratory Network (LabNet) regularly issues international proficiency studies. The 2010 HPV genotyping proficiency panel for HPV vaccinology contained 43 coded samples composed of purified plasmids of 16 HPV types (HPV types 6, 11, 16, 18, 31, 33, 35, 39, 45, 51, 52, 56, 58, 59, 66, and 68a and 68b) and 3 coded extraction controls. Proficient typing was defined as detection in both single and multiple infections of 50 international units (IU) of HPV type 16 (HPV-16) and HPV-18 DNA and 500 genome equivalents (GE) for the other 14 HPV types. Ninety-eight laboratories worldwide submitted a total of 132 data sets. Twenty-four different HPV genotyping assay methods were used, with Linear Array being the most commonly used. Other major assays used were a line blot assay (Inno-LiPa), CLART, type-specific real-time PCR, PCR Luminex, and different microarray assays. Altogether, 72 data sets were proficient for detection of more than 1 type, and only 26 data sets proficiently detected all 16 HPV types. The major oncogenic HPV types, 16 and 18, were proficiently detected in 95.0% (114/120) and 87.0% (94/108) of data sets, respectively. Forty-six data sets reported multiple false-positive results and were considered nonproficient. A trend toward increased sensitivity of assays was seen for the 41 laboratories that participated in both 2008 and 2010. In conclusion, continued global proficiency studies will be required for establishing comparable and reliable HPV genotyping services for vaccinology worldwide.