MilliporeSigma
Reduced exercise endurance in interleukin-6-deficient mice.
Endocrinology (2004-02-28)
Jenny Fäldt, Ingrid Wernstedt, Sharyn M Fitzgerald, Kristina Wallenius, Göran Bergström, John-Olov Jansson
ABSTRACT
IL-6 is produced and released in large amounts from skeletal muscle during prolonged exercise in both mice and humans, but there are few data indicating the biological significance of this. IL-6 exerts metabolic effects such as stimulating energy expenditure and reducing body fat mass. We have now investigated the effects of IL-6 deficiency on exercise endurance and energy expenditure in preobese and obese IL-6-deficient (IL-6(-/-)) mice. Four-month-old preobese and 7-month-old obese IL-6(-/-) male mice backcrossed to C57BL/6 and their littermate controls were exercised on a treadmill, and energy expenditure was measured as oxygen consumption with the use of indirect calorimetry. The preobese IL-6(-/-) mice were significantly leaner than the control mice, whereas the older IL-6(-/-) mice, as expected, had developed obesity. Resting young, but not older, IL-6(-/-) mice had an elevated respiratory exchange ratio (RER), indicating that they oxidize carbohydrates rather than fat for energy utilization. During exercise, the young and older IL-6(-/-) mice had a reduced endurance and a progressive decrease in oxygen consumption compared with control mice. There was no difference in RER in young IL-6(-/-) mice, whereas RER was enhanced in older IL-6(-/-), mice during exercise. In summary, IL-6(-/-) mice have reduced endurance and energy expenditure during exercise, suggesting that IL-6 is necessary for normal exercise capacity.
Product Description
Supelco
Glucose Standard Solution, 1 mg/mL
Pricing and availability is not currently available.