MilliporeSigma
Fibrosis on a Chip for Screening of Anti-Fibrosis Drugs.
Methods in molecular biology (Clifton, N.J.) (2021-05-25)
Mohammadnabi Asmani, Ruogang Zhao
ABSTRACT
Idiopathic pulmonary fibrosis (IPF) is a chronic pathological disorder that targets alveoli interstitial tissues and is characterized by the progressive stiffening of alveolar membrane. The median survival rate of the patients with IPF is less than 5 years. Currently, IPF has no cure and there are few options to alleviate the progress of this disease. A critical roadblock in developing new anti-fibrosis therapies is the absence of reliable cell based in vitro models that can recapitulate the progressive features of this disease. Here a novel fibrotic microtissue on a chip system is created to model the fibrotic transition of the lung interstitial tissue and the effect of anti-fibrosis drugs on such transitions. This system will not only help to expedite the efficacy analysis of anti-fibrotic therapies but also help to unveil their potential mode of action.
Sigma-Aldrich
Anti-Rat Collagen Type I Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Procollagen Type I Antibody, CT, clone PCIDG10 (Ascites Free), clone PCIDG10, from mouse
Pricing and availability is not currently available.