MilliporeSigma
  • A deubiquitination module essential for Treg fitness in the tumor microenvironment.
A deubiquitination module essential for Treg fitness in the tumor microenvironment.
Science advances (2022-11-26)
Elena Montauti, Samuel E Weinberg, Peng Chu, Shuvam Chaudhuri, Nikita L Mani, Radhika Iyer, Yuanzhang Zhou, Yusi Zhang, Changhong Liu, Chen Xin, Shana Gregory, Juncheng Wei, Yana Zhang, Wantao Chen, Zhaolin Sun, Ming Yan, Deyu Fang
ABSTRACT
The tumor microenvironment (TME) enhances regulatory T (Treg) cell stability and immunosuppressive functions through up-regulation of lineage transcription factor Foxp3, a phenomenon known as Treg fitness or adaptation. Here, we characterize previously unknown TME-specific cellular and molecular mechanisms underlying Treg fitness. We demonstrate that TME-specific stressors including transforming growth factor-β (TGF-β), hypoxia, and nutrient deprivation selectively induce two Foxp3-specific deubiquitinases, ubiquitin-specific peptidase 22 (Usp22) and Usp21, by regulating TGF-β, HIF, and mTOR signaling, respectively, to maintain Treg fitness. Simultaneous deletion of both USPs in Treg cells largely diminishes TME-induced Foxp3 up-regulation, alters Treg metabolic signatures, impairs Treg-suppressive function, and alleviates Treg suppression on cytotoxic CD8+ T cells. Furthermore, we developed the first Usp22-specific small-molecule inhibitor, which dramatically reduced intratumoral Treg Foxp3 expression and consequently enhanced antitumor immunity. Our findings unveil previously unappreciated mechanisms underlying Treg fitness and identify Usp22 as an antitumor therapeutic target that inhibits Treg adaptability in the TME.
Sigma-Aldrich
mTOR Inhibitor XI, Torin1, mTOR Inhibitor XI, Torin1, CAS 1222998-36-8, is a cell-permeable, highly potent, ATP-competitive inhibitor of mTOR and DNA-PK (IC50 = 4.32 and 6.34 nM, respectively).
Pricing and availability is not currently available.
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Usp22i-S02, ≥98% (HPLC)
Pricing and availability is not currently available.