MilliporeSigma
  • Protein interaction networks in the vasculature prioritize genes and pathways underlying coronary artery disease.
Protein interaction networks in the vasculature prioritize genes and pathways underlying coronary artery disease.
Communications biology (2024-01-13)
Qiuyu Martin Zhu, Yu-Han H Hsu, Frederik H Lassen, Bryan T MacDonald, Stephanie Stead, Edyta Malolepsza, April Kim, Taibo Li, Taiji Mizoguchi, Monica Schenone, Gaelen Guzman, Benjamin Tanenbaum, Nadine Fornelos, Steven A Carr, Rajat M Gupta, Patrick T Ellinor, Kasper Lage
ABSTRACT
Population-based association studies have identified many genetic risk loci for coronary artery disease (CAD), but it is often unclear how genes within these loci are linked to CAD. Here, we perform interaction proteomics for 11 CAD-risk genes to map their protein-protein interactions (PPIs) in human vascular cells and elucidate their roles in CAD. The resulting PPI networks contain interactions that are outside of known biology in the vasculature and are enriched for genes involved in immunity-related and arterial-wall-specific mechanisms. Several PPI networks derived from smooth muscle cells are significantly enriched for genetic variants associated with CAD and related vascular phenotypes. Furthermore, the networks identify 61 genes that are found in genetic loci associated with risk of CAD, prioritizing them as the causal candidates within these loci. These findings indicate that the PPI networks we have generated are a rich resource for guiding future research into the molecular pathogenesis of CAD.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-PDIA6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-KIAA1462 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution