MilliporeSigma
  • Evaluation of apoptosis induced by nanoparticles and fine particles in RAW 264.7 macrophages: facts and artefacts.
Evaluation of apoptosis induced by nanoparticles and fine particles in RAW 264.7 macrophages: facts and artefacts.
Toxicology in vitro : an international journal published in association with BIBRA (2011-12-27)
Verena Wilhelmi, Ute Fischer, Damiën van Berlo, Klaus Schulze-Osthoff, Roel P F Schins, Catrin Albrecht
ABSTRACT
Current hazard characterisation of nanoparticles (NP) is predominantly based on in vitro test systems, being established for small molecules of drugs and chemicals. However, specific physicochemical properties of NP may result in interference with assay components, biomarkers, or detection systems. In the present study, six types of (nano)particles were screened in RAW 264.7 macrophages by common cytotoxicity methods (WST-1, LDH). Our specific focus was on the investigation of apoptosis (analysis of hypodiploid DNA, phosphatidylserine exposure, caspase 3/7 activation, and Cell Death Detection ELISA). Assays were validated by the well-known apoptosis inducer staurosporine. Our results show that ZnO, DQ12 quartz and amorphous silica are cytotoxic with strong indications for apoptotic effects in RAW 264.7 macrophages, whereas toxicity was absent for MgO. For fine as well as ultrafine TiO(2), no apoptotic effects could be detected except for induction of DNA fragmentation. The results of our study demonstrate the necessity to control on a case-by-case basis for assay interference to avoid misinterpretation of specific in vitro test findings. To obtain valid statements on the potential induction of apoptosis by specific NP the measurement of multiple endpoints is a prerequisite.
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Pricing and availability is not currently available.