MilliporeSigma
  • Ferrocene-Mediated carbon paste electrode modified with D-fructose dehydrogenase for batch mode measurement of D-fructose.
Ferrocene-Mediated carbon paste electrode modified with D-fructose dehydrogenase for batch mode measurement of D-fructose.
Applied biochemistry and biotechnology (2000-11-09)
M Boujtita, N El Murr
ABSTRACT
A mediated modified carbon paste and renewable surface electrode for fructose amperometric measurement based on D-fructose dehydrogenase (FDH) was prepared and optimized. Commercially available ferrocene (FcH) and hydroxymethyl ferrocene (FcCH2OH) were used as mediators. The substituted FcH showed better linearity and higher sensitivity. The influence of different experimental parameters was studied for optimum analytical performance. The final FDH-modified electrode showed good analytical performance for batch mode measurements of fructose.