MilliporeSigma
  • Evaluation of the information content of RNA structure mapping data for secondary structure prediction.
Evaluation of the information content of RNA structure mapping data for secondary structure prediction.
RNA (New York, N.Y.) (2010-04-24)
Scott Quarrier, Joshua S Martin, Lauren Davis-Neulander, Arthur Beauregard, Alain Laederach
ABSTRACT
Structure mapping experiments (using probes such as dimethyl sulfate [DMS], kethoxal, and T1 and V1 RNases) are used to determine the secondary structures of RNA molecules. The process is iterative, combining the results of several probes with constrained minimum free-energy calculations to produce a model of the structure. We aim to evaluate whether particular probes provide more structural information, and specifically, how noise in the data affects the predictions. Our approach involves generating "decoy" RNA structures (using the sFold Boltzmann sampling procedure) and evaluating whether we are able to identify the correct structure from this ensemble of structures. We show that with perfect information, we are always able to identify the optimal structure for five RNAs of known structure. We then collected orthogonal structure mapping data (DMS and RNase T1 digest) under several solution conditions using our high-throughput capillary automated footprinting analysis (CAFA) technique on two group I introns of known structure. Analysis of these data reveals the error rates in the data under optimal (low salt) and suboptimal solution conditions (high MgCl(2)). We show that despite these errors, our computational approach is less sensitive to experimental noise than traditional constraint-based structure prediction algorithms. Finally, we propose a novel approach for visualizing the interaction of chemical and enzymatic mapping data with RNA structure. We project the data onto the first two dimensions of a multidimensional scaling of the sFold-generated decoy structures. We are able to directly visualize the structural information content of structure mapping data and reconcile multiple data sets.
Sigma-Aldrich
Ribonuclease T1 from Aspergillus oryzae, ammonium sulfate suspension, 300,000-600,000 units/mg protein
Pricing and availability is not currently available.