MilliporeSigma
  • Highly efficient chemoenzymatic synthesis of beta1-4-linked galactosides with promiscuous bacterial beta1-4-galactosyltransferases.
Highly efficient chemoenzymatic synthesis of beta1-4-linked galactosides with promiscuous bacterial beta1-4-galactosyltransferases.
Chemical communications (Cambridge, England) (2010-07-14)
Kam Lau, Vireak Thon, Hai Yu, Li Ding, Yi Chen, Musleh M Muthana, Denton Wong, Ronald Huang, Xi Chen
ABSTRACT
Two bacterial beta1-4-galactosyltransferases, NmLgtB and Hp1-4GalT, exhibit promiscuous and complementary acceptor substrate specificity. They have been used in an efficient one-pot multienzyme system to synthesize LacNAc, lactose, and their derivatives including those containing negatively charged 6-O-sulfated GlcNAc and C2-substituted GlcNAc or Glc, from monosaccharide derivatives and inexpensive Glc-1-P.