MilliporeSigma
Advances in non-snake venom L-amino acid oxidase.
Applied biochemistry and biotechnology (2012-03-01)
Zhiliang Yu, Hua Qiao
ABSTRACT
L-amino acid oxidase is widely found in diverse organisms and has different properties. It is thought to contribute to antimicrobial activity, amino acid catabolism, and so forth. The purpose of this communication is to summarize the advances in non-snake venom L-amino acid oxidase, including its enzymatic and structural properties, gene cloning and expression, and biological function. In addition, the mechanism of its biological function as well as its application is also discussed.
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus adamanteus, Type I (dried venom)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus atrox (Western Diamondback Rattlesnake), Type VI, dried venom
Pricing and availability is not currently available.
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus adamanteus, Type IV, ≥4.0 units/mg protein, aqueous suspension
Pricing and availability is not currently available.